This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01617425_g1
See other PGAM1 GE Assays ›
Gene Symbol:
Pgam1
Gene Name:
phosphoglycerate mutase 1
Gene Aliases:
2310050F24Rik, Pgam-1
Chromosome Location:
Chr.19: 41911871 - 41918665 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01617425_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pgam1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023418.2 NP_075907.2 1 - 2 318 114
GenBank mRNA AF283667.1 - 1 - 2 230 114
AK004921.1 - 1 - 2 174 114
AK009905.2 - 1 - 2 171 114
AK144681.1 - 1 - 2 172 115
AK150716.1 - 1 - 2 167 114
AK152443.1 - 1 - 2 167 114
AK153202.1 - 1 - 2 171 114
AK159305.1 - 1 - 2 171 114
AK165410.1 - 1 - 2 167 114
AK168949.1 - 1 - 2 164 114
AK194151.1 - 1 - 2 81 114
BC002241.1 - 1 - 2 266 3491964 114
BC005661.1 - 1 - 2 232 3588761 114
BC066844.1 - 1 - 2 155 30471755 114
BC083090.1 - 1 - 2 167 30444229 114
BC106139.1 - 1 - 2 238 6528521 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pgam1

Panther Classification:

Molecular Function -

mutase

Gene Ontology Categories:

Function(s) Process(es)

glycolytic process
regulation of glycolytic process
metabolic process
regulation of pentose-phosphate shunt
respiratory burst
catalytic activity
bisphosphoglycerate mutase activity
bisphosphoglycerate 2-phosphatase activity
phosphoglycerate mutase activity
hydrolase activity
isomerase activity
intramolecular transferase activity, phosphotransferases
protein kinase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0