This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01610895_g1
See other HDAC1 GE Assays ›
Gene Symbol:
Hdac1
Gene Name:
histone deacetylase 1
Gene Aliases:
HD1, Hdac1-ps, MommeD5, RPD3
Chromosome Location:
Chr.4: 129516104 - 129542646 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01610895_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hdac1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008228.2 NP_032254.1 6 - 7 652 70
GenBank mRNA AK076080.1 - 6 - 7 637 70
BC092070.1 - 6 - 7 647 4217199 70
BC092285.1 - 6 - 7 652 70
BC108371.1 - 6 - 7 653 4976514 70
U80780.1 - 6 - 7 649 70
X98207.1 - 6 - 7 652 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hdac1

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
response to amphetamine
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
protein deacetylation
endoderm development
circadian rhythm
positive regulation of cell proliferation
negative regulation of cell proliferation
epidermal cell differentiation
negative regulation of gene expression
positive regulation of receptor biosynthetic process
chromatin modification
histone deacetylation
hippocampus development
neuron differentiation
positive regulation of interleukin-1 production
positive regulation of tumor necrosis factor production
circadian regulation of gene expression
cellular response to oxidative stress
odontogenesis of dentin-containing tooth
positive regulation of tyrosine phosphorylation of Stat3 protein
embryonic digit morphogenesis
ATP-dependent chromatin remodeling
negative regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of neuron apoptotic process
negative regulation by host of viral transcription
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of insulin secretion
rhythmic process
positive regulation of oligodendrocyte differentiation
negative regulation of androgen receptor signaling pathway
hair follicle placode formation
eyelid development in camera-type eye
fungiform papilla formation
histone H3 deacetylation
histone H4 deacetylation
negative regulation of canonical Wnt signaling pathway
positive regulation of chemokine (C-X-C motif) ligand 2 production
positive regulation of type B pancreatic cell apoptotic process
negative regulation of peptidyl-lysine acetylation
negative regulation of intrinsic apoptotic signaling pathway
transcription regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
core promoter binding
RNA polymerase II repressing transcription factor binding
RNA polymerase II transcription corepressor activity
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
histone deacetylase activity
protein binding
transcription factor binding
hydrolase activity
deacetylase activity
enzyme binding
histone deacetylase activity (H3-K14 specific)
nucleosomal DNA binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
histone deacetylase activity (H3-K9 specific)
protein complex binding
protein deacetylase activity
activating transcription factor binding
histone deacetylase activity (H4-K16 specific)
Krueppel-associated box domain binding
histone deacetylase binding
transcription regulatory region DNA binding
NAD-dependent histone deacetylase activity (H3-K9 specific)
NAD-dependent histone deacetylase activity (H4-K16 specific)
protein N-terminus binding
NF-kappaB binding
repressing transcription factor binding
NAD-dependent histone deacetylase activity (H3-K18 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0