This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01354528_g1
See other SRPX2 GE Assays ›
Gene Symbol:
Srpx2
Gene Name:
sushi-repeat-containing protein, X-linked 2
Gene Aliases:
1110039C07Rik, SRP, SRPUL
Chromosome Location:
Chr.X: 133908416 - 133932448 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01354528_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srpx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001083895.3 NP_001077364.2 5 - 6 1026 110
NM_026838.4 NP_081114.2 6 - 7 1078 110
GenBank mRNA AF393640.1 - 4 - 5 596 110
AK136935.1 - 5 - 6 881 110
BC028307.1 - 6 - 7 807 3488526 110
BQ830905.1 - 2 - 3 237 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001271.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srpx2

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
cell adhesion
single organismal cell-cell adhesion
regulation of phosphorylation
cell motility
positive regulation of synapse assembly
vocalization behavior
positive regulation of cell migration involved in sprouting angiogenesis
receptor binding
hepatocyte growth factor binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00007835.mm.1 NM_001083895.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007835.mm.1 NM_026838.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007835.mm.1 TC0X00001271.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007836.mm.1 NM_001083895.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007836.mm.1 NM_026838.4 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007836.mm.1 TC0X00001271.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0