This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01354448_m1
See other PADI3 GE Assays ›
Gene Symbol:
Padi3
Gene Name:
peptidyl arginine deiminase, type III
Gene Aliases:
AI507004, Pad3, Pdi3, mKIAA4171
Chromosome Location:
Chr.4: 140785369 - 140810648 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01354448_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Padi3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011060.4 NP_035190.3 3 - 4 389 65
GenBank mRNA AB013849.1 - 3 - 4 390 65
AK220522.1 - 2 - 3 631 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003866.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Padi3

Gene Ontology Categories:

Function(s) Process(es)

protein citrullination
protein-arginine deiminase activity
calcium ion binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400031919.mm.1 NM_011060.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400031919.mm.1 NM_011060.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400031919.mm.1 TC0400003866.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0