This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01351094_g1
See other PSD3 GE Assays ›
Gene Symbol:
Psd3
Gene Name:
pleckstrin and Sec7 domain containing 3
Gene Aliases:
4931420C21Rik, AI661273, BC003498, D430018P08, EFA6D
Chromosome Location:
Chr.8: 67689082 - 68206052 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01351094_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Psd3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030263.5 NP_084539.2 2 - 3 343 102
NM_177698.4 NP_808366.2 4 - 5 2104 102
GenBank mRNA AB220685.1 - 4 - 5 2104 102
AK016466.1 - 2 - 3 398 102
AK028684.1 - 5 - 6 1916 102
AK161498.1 - 2 - 3 329 102
BC052365.1 - 2 - 3 307 102
CJ047427.1 - 2 - 3 344 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002339.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Psd3

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

neuron differentiation
ARF protein signal transduction
regulation of ARF protein signal transduction
positive regulation of GTPase activity
guanyl-nucleotide exchange factor activity
ARF guanyl-nucleotide exchange factor activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800017605.mm.1 NM_030263.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017605.mm.1 NM_030263.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800017605.mm.1 NM_177698.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017605.mm.1 TC0800002339.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800017606.mm.1 NM_030263.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017606.mm.1 NM_030263.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800017606.mm.1 TC0800002339.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800017604.mm.1 NM_030263.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017604.mm.1 NM_030263.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800017604.mm.1 NM_177698.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800017604.mm.1 TC0800002339.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 695892 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0