Array Name |
Probe IDs |
Targets |
Target Type |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
284843 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
AF211854.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
AF211855.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
BC036323.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
BC051061.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
NM_133648.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920753 |
NM_133649.2 |
refseq_mrna_mouse_NM |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P131766 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943788 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943878 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943783 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943833 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943888 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943892 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943796 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943886 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943834 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03592548 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943877 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03379201 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943828 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03426163 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943795 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03625745 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943896 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943876 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03820826 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943779 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943805 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943810 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943831 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943786 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03734041 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943775 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943897 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943813 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03734037 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03442187 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943794 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943899 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943881 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943778 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03820822 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03786041 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943901 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943780 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943882 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03302142 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943894 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943821 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943785 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943905 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943800 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943776 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943903 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03786807 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03786809 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P03786816 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P22943804 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P04045586 |
- |
- |