This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350911_g1
See other 4930556C24RIK GE Assays ›
Gene Symbol:
4930556C24Rik
Gene Name:
RIKEN cDNA 4930556C24 gene
Gene Aliases:
-
Chromosome Location:
Chr.16: 85819211 - 85826772 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350911_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930556C24Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040506.1 2 - 3 265 65
GenBank mRNA AK016142.1 - 2 - 3 266 65
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715748.1 - 2 - 3 266 Chr. 16 - 85819211 - 85826784 [+] on Build GRCm38 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif,
1
Adamts1 ADAM-TS1,
ADAMTS,
ADAMTS-1,
C3-C5,
METH-1,
METH1
-22.1kb Anti-sense Chr. 16 - 85793827 - 85803115 on NCBI Build GRCm38
Downstream a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif,
5 (aggrecanase-2)
Adamts5 9530092O11Rik,
ADAM-TS5,
ADAMTS1,
ADAMTS11,
AI481094,
ASMP-2
32.6kb Anti-sense Chr. 16 - 85858157 - 85902374 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000946.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600007396.mm.1 NR_040506.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007396.mm.1 NR_040506.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600007396.mm.1 TC1600000946.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0