This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350255_m1
See other MOB1B GE Assays ›
Gene Symbol:
Mob1b
Gene Name:
MOB kinase activator 1B
Gene Aliases:
1110003E08Rik, AU015450, B230364F10, Mobkl1a
Chromosome Location:
Chr.5: 88720871 - 88758455 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350255_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mob1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026735.2 NP_081011.1 3 - 4 511 62
GenBank mRNA AK003346.2 - 3 - 4 406 62
AK077320.1 - 3 - 4 487 62
AK150016.1 - 3 - 4 504 62
AK162120.1 - 3 - 4 511 62
AK172291.1 - 3 - 4 479 62
BC104402.1 - 4 - 5 342 40050502 62
BC104403.1 - 3 - 4 406 40050503 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000815.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mob1b

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

hippo signaling
positive regulation of phosphorylation
positive regulation of catalytic activity
protein autophosphorylation
kinase activator activity
kinase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500007197.mm.1 NM_026735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007197.mm.1 NM_026735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500007197.mm.1 TC0500000815.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500007193.mm.1 NM_026735.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007193.mm.1 NM_026735.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500007193.mm.1 TC0500000815.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0