This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01350197_m1
See other MELK GE Assays ›
Gene Symbol:
Melk
Gene Name:
maternal embryonic leucine zipper kinase
Gene Aliases:
AI327312, MPK38, mKIAA0175
Chromosome Location:
Chr.4: 44300876 - 44364675 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01350197_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Melk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010790.2 NP_034920.2 10 - 11 1033 62
GenBank mRNA AK010498.1 - 10 - 11 999 62
AK011932.1 - 10 - 11 1033 62
AK129076.1 - 10 - 11 986 62
AK137000.1 - 10 - 11 1059 62
AK145021.1 - 10 - 11 968 62
AK145316.1 - 10 - 11 942 62
AK161029.1 - 10 - 11 1042 62
AK164138.1 - 10 - 11 1000 62
AK167457.1 - 10 - 11 957 62
BC020416.1 - 9 - 10 868 62
BC085276.1 - 10 - 11 1066 30443236 62
L76158.1 - 10 - 11 964 62
X95351.1 - 10 - 11 971 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Melk

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
cell cycle
cell proliferation
intrinsic apoptotic signaling pathway in response to oxidative stress
phosphorylation
peptidyl-tyrosine phosphorylation
positive regulation of apoptotic process
protein autophosphorylation
neural precursor cell proliferation
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
non-membrane spanning protein tyrosine kinase activity
calcium ion binding
protein binding
ATP binding
lipid binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0