This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01349916_m1
See other DGKG GE Assays ›
Gene Symbol:
Dgkg
Gene Name:
diacylglycerol kinase, gamma
Gene Aliases:
2900055E17Rik, 90kDa, AI854428, Dagk3, E430001K23Rik, mKIAA4131
Chromosome Location:
Chr.16: 22466568 - 22657299 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01349916_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgkg
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138650.2 NP_619591.1 23 - 24 2666 65
GenBank mRNA AK083258.1 - 22 - 23 2542 65
AK220294.1 - 23 - 24 2608 65
BC015278.1 - 22 - 23 2239 4221290 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001380.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgkg

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
intracellular signal transduction
nucleotide binding
diacylglycerol kinase activity
calcium ion binding
ATP binding
kinase activity
transferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600011477.mm.1 NM_138650.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600011477.mm.1 NM_138650.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600011477.mm.1 TC1600001380.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0