This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01348257_m1
See other PDE9A GE Assays ›
Gene Symbol:
Pde9a
Gene Name:
phosphodiesterase 9A
Gene Aliases:
PDE9A1
Chromosome Location:
Chr.17: 31386212 - 31476309 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01348257_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde9a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163748.1 NP_001157220.1 15 - 16 1333 66
NM_008804.4 NP_032830.3 15 - 16 1333 66
GenBank mRNA AF031147.1 - 15 - 16 1288 66
AF068247.1 - 15 - 16 1297 66
AK030509.1 - 15 - 16 1317 66
AK168143.1 - 15 - 16 1315 66
BC025882.1 - 14 - 15 1464 66
BC038675.1 - 15 - 16 1282 66
BC046915.1 - 15 - 16 1282 66
BC061163.1 - 15 - 16 1313 6533665 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000535.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde9a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
3',5'-cyclic-nucleotide phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700004912.mm.1 NM_001163748.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004912.mm.1 NM_001163748.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004912.mm.1 NM_008804.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700004912.mm.1 NM_008804.4 Refseq
Affymetrix Mouse Clariom S Array PSR1700004912.mm.1 TC1700000535.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0