This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01347731_m1
See other DIAP2 GE Assays ›
Gene Symbol:
Diap2
Gene Name:
diaphanous homolog 2 (Drosophila)
Gene Aliases:
Dia3, Diaph2, Drf2, E430022I22Rik
Chromosome Location:
Chr.X: 129749668 - 130465834 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01347731_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Diap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172493.2 NP_766081.1 14 - 15 1745 127
GenBank mRNA AB713187.1 - 15 - 16 1540 127
AK083254.1 - 7 - 8 677 127
AK088648.1 - 14 - 15 1679 127
AK153986.1 - 14 - 15 1733 127
AY312280.1 - 14 - 15 1682 127

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00001254.mm.1

Set Membership:

Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
multicellular organism development
female gamete generation
cellular component organization
actin cytoskeleton organization
actin filament polymerization
cell differentiation
oogenesis
actin binding
Rho GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00007772.mm.1 NM_172493.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007772.mm.1 NM_172493.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007772.mm.1 TC0X00001254.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0X00007773.mm.1 NM_172493.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007773.mm.1 NM_172493.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00007773.mm.1 TC0X00001254.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0