This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01347648_m1
See other FAM47E GE Assays ›
Gene Symbol:
Fam47e
Gene Name:
family with sequence similarity 47, member E
Gene Aliases:
C730043O17, Gm1381
Chromosome Location:
Chr.5: 92554951 - 92591285 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01347648_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam47e
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170571.1 NP_001164042.1 2 - 3 319 81
GenBank mRNA AA244788.1 - 2 - 3 300 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000876.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500007622.mm.1 NM_001033478.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007622.mm.1 NM_001033478.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500007622.mm.1 NM_001170571.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007622.mm.1 NM_001170571.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500007622.mm.1 TC0500000876.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0