This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01347581_m1
See other SEPT11 GE Assays ›
Gene Symbol:
Sept11
Gene Name:
septin 11
Gene Aliases:
6230410I01Rik, AW548875, D5Ertd606e
Chromosome Location:
Chr.5: 93091879 - 93179054 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01347581_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sept11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009818.1 NP_001009818.1 3 - 4 450 71
GenBank mRNA AK028475.1 - 3 - 4 382 71
AK136063.1 - 1 - 2 79 71
AK166517.1 - 3 - 4 471 71
AK170945.1 - 2 - 3 260 71
AK171368.1 - 3 - 4 471 71
BC031456.1 - 3 - 4 357 4037515 71
BC062206.1 - 3 - 4 491 5150858 71
BC064466.1 - 3 - 4 450 6825549 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000888.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sept11

Panther Classification:

Molecular Function -

cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein heterooligomerization
cell division
nucleotide binding
molecular_function
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500007696.mm.1 NM_001009818.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500007696.mm.1 TC0500000888.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0