Assay Details
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143610.1 | - | 11 - 12 | 3786 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143610.1 | - | 11 - 12 | 3786 | 69 |
Set Membership: |
![]() ![]() ![]() |