This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01345148_m1
See other PPEF2 GE Assays ›
Gene Symbol:
Ppef2
Gene Name:
protein phosphatase, EF hand calcium-binding domain 2
Gene Aliases:
-
Chromosome Location:
Chr.5: 92226691 - 92256900 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01345148_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppef2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011148.3 NP_035278.1 10 - 11 1332 97
GenBank mRNA AF023458.1 - 10 - 11 1332 97
BC027049.1 - 11 - 12 1639 5356685 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002752.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppef2

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

visual perception
negative regulation of peptidyl-threonine phosphorylation
dephosphorylation
negative regulation of MAPK cascade
response to stimulus
detection of stimulus involved in sensory perception
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
iron ion binding
calcium ion binding
hydrolase activity
manganese ion binding
Hsp70 protein binding
mitogen-activated protein kinase kinase kinase binding
metal ion binding
Hsp90 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500024234.mm.1 NM_011148.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500024234.mm.1 NM_011148.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500024234.mm.1 TC0500002752.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0