Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600010516.mm.1 |
NM_001145435.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010516.mm.1 |
NM_001145435.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010516.mm.1 |
NM_029053.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010516.mm.1 |
NM_029053.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010516.mm.1 |
TC1600001293.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600010514.mm.1 |
NM_029053.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010514.mm.1 |
NM_029053.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600010514.mm.1 |
TC1600001293.mm.1 |
MTA TC |