This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344190_m1
See other FILIP1L GE Assays ›
Gene Symbol:
Filip1l
Gene Name:
filamin A interacting protein 1-like
Gene Aliases:
4631422O05Rik, Doc1
Chromosome Location:
Chr.16: 57353277 - 57572804 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344190_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Filip1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177871.1 NP_001171342.1 1 - 2 94 99
GenBank mRNA AK019472.2 - 1 - 2 95 99
AK132319.1 - 1 - 2 94 99
BC024523.1 - 1 - 2 86 99
BC062166.1 - 1 - 2 77 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001177871.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2109594 AK146705.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC098507.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC062166.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 AK132319.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC037751.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 NM_001177871.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC098507.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC062166.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 AK132319.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 BC037751.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P142153 NM_001177871.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2109594 AK146705.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134569 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10436370 - -
Affymetrix Mouse Gene 1.0 ST Array 10436365 - -
Affymetrix Mouse Gene 1.0 ST Array 10436371 - -
Affymetrix Mouse Gene 1.0 ST Array 10436367 - -
Affymetrix Mouse Gene 1.0 ST Array 10436366 - -
Affymetrix HT MG-430 PM Array Plate 1428861_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10436369 - -
Affymetrix Mouse Gene 1.1 ST Array 10436363 - -
Mouse Genome 430 2.0 Array 1446296_PM_at - -
Mouse Genome 430 2.0 Array 1428861_at - -
Mouse Genome 430 2.0 Array 1446296_at - -
Mouse Genome 430 2.0 Array 1428861_PM_at - -
Affymetrix Mouse Clariom S Array PSR1600006566.mm.1 NM_001040397.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006566.mm.1 NM_001040397.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600006566.mm.1 NM_001177871.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006566.mm.1 NM_001177871.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006566.mm.1 TC1600000756.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600006565.mm.1 NM_001177871.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006565.mm.1 NM_001177871.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600006565.mm.1 TC1600000756.mm.1 MTA TC
Mouse Expression Set 430 1428861_at - -
Mouse Expression Set 430 1428861_PM_at - -
Murine Genome U74v2 Set 166775_PM_at - -
Murine Genome U74v2 Set 166775_at - -
Affymetrix HT Mouse Genome 430B Array 1428861_at - -
Affymetrix HT Mouse Genome 430B Array 1428861_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 396120 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 103360541 - -
Illumina mouse-6 v1.0 expression beadchip 103360541 - -
mouse-6 v1.1 (Illumina) scl0001851.1_197-S - -
Illumina Sentrix mouse-6.v1 bead array 103360541 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 103360541 - -
MouseRef-8 v2 Expression BeadChip ILMN_2817473 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2817473 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2455575 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0