This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01344167_g1
See other ADAMTS1 GE Assays ›
Gene Symbol:
Adamts1
Gene Name:
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 1
Gene Aliases:
ADAM-TS1, ADAMTS, ADAMTS-1, C3-C5, METH-1, METH1
Chromosome Location:
Chr.16: 85793827 - 85803115 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01344167_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adamts1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009621.4 NP_033751.3 5 - 6 2072 59
GenBank mRNA AK156830.1 - 5 - 6 2071 59
AK161409.1 - 5 - 6 1174 59
AK163622.1 - 5 - 6 2071 59
BC003269.1 - 5 - 6 2057 59
BC040382.1 - 5 - 6 2056 3156736 59
BC050834.1 - 5 - 6 2056 6505494 59
D67076.1 - 5 - 6 2056 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adamts1

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

ovulation from ovarian follicle
kidney development
proteolysis
positive regulation of neuron projection development
negative regulation of angiogenesis
heart trabecula formation
metalloendopeptidase activity
protein binding
heparin binding
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0