Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600004989.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004989.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004989.mm.1 |
TC1600000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004985.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004985.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004985.mm.1 |
TC1600000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004990.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004990.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004990.mm.1 |
TC1600000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004986.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004986.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004986.mm.1 |
TC1600000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004987.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004987.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004987.mm.1 |
TC1600000529.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600004988.mm.1 |
NM_008225.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004988.mm.1 |
NM_008225.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600004988.mm.1 |
TC1600000529.mm.1 |
MTA TC |