This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343524_g1
See other HCLS1 GE Assays ›
Gene Symbol:
Hcls1
Gene Name:
hematopoietic cell specific Lyn substrate 1
Gene Aliases:
AW213261, HS1
Chromosome Location:
Chr.16: 36934983 - 36963214 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343524_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hcls1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008225.2 NP_032251.2 12 - 13 1356 103
GenBank mRNA AK088723.1 - 12 - 13 1338 103
AK132608.1 - 3 - 4 404 103
BC007469.1 - 12 - 13 1326 3593456 103
CA321698.1 - 3 - 4 438 103
D42120.1 - 12 - 13 1287 103
X84797.1 - 12 - 13 1324 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000529.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hcls1

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein cytoskeletal protein non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
positive regulation of cell proliferation
response to hormone
positive regulation of phosphatidylinositol 3-kinase signaling
actin filament polymerization
erythrocyte differentiation
regulation of actin filament polymerization
positive regulation of granulocyte differentiation
positive regulation of peptidyl-serine phosphorylation
positive regulation of tyrosine phosphorylation of STAT protein
positive regulation of transcription factor import into nucleus
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to cytokine stimulus
negative regulation of leukocyte apoptotic process
RNA polymerase II transcription factor binding
actin binding
protein binding
SH3 domain binding
protein kinase binding
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005001.mm.1 NM_008225.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005001.mm.1 NM_008225.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600005001.mm.1 TC1600000529.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0