This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343507_g1
See other SLC15A2 GE Assays ›
Gene Symbol:
Slc15a2
Gene Name:
solute carrier family 15 (H+/peptide transporter), member 2
Gene Aliases:
8430408C16Rik, C78862, Pept2
Chromosome Location:
Chr.16: 36750164 - 36785157 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343507_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc15a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021301.3 NP_067276.2 21 - 22 2216 64
GenBank mRNA AF111811.1 - 21 - 22 2216 64
AK128939.1 - 21 - 22 2049 64
BC138627.1 - 21 - 22 2173 8861647 64
BC138628.1 - 21 - 22 2173 8861648 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001582.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc15a2

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

oligopeptide transport
antibiotic transport
dipeptide transport
high-affinity oligopeptide transporter activity
antibiotic transporter activity
dipeptide transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600012895.mm.1 NM_021301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600012895.mm.1 NM_021301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600012895.mm.1 TC1600001582.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600012897.mm.1 NM_021301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600012897.mm.1 NM_021301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600012897.mm.1 TC1600001582.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600012894.mm.1 NM_021301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600012894.mm.1 NM_021301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600012894.mm.1 TC1600001582.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600012896.mm.1 NM_021301.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600012896.mm.1 NM_021301.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600012896.mm.1 TC1600001582.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0