This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01343076_g1
See other HEG1 GE Assays ›
Gene Symbol:
Heg1
Gene Name:
HEG homolog 1 (zebrafish)
Gene Aliases:
4632417D23Rik, 5530401I02Rik, 9530025L16Rik, BC038614, Gm629, OTTMUSG00000027315
Chromosome Location:
Chr.16: 33684430 - 33771576 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01343076_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Heg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175256.5 NP_780465.4 8 - 9 2607 78
GenBank mRNA BC076596.1 - 6 - 7 1886 30645937 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000475.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Heg1

Gene Ontology Categories:

Function(s) Process(es)

vasculogenesis
in utero embryonic development
endothelial cell development
endothelial cell morphogenesis
vasculature development
lymph vessel development
lymph circulation
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular septum development
cell-cell junction assembly
multicellular organism development
heart development
post-embryonic development
lung development
multicellular organism growth
cell-cell junction organization
venous blood vessel morphogenesis
regulation of body fluid levels
cardiac muscle tissue growth
pericardium development
positive regulation of fibroblast growth factor production
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600004503.mm.1 NM_175256.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004503.mm.1 NM_175256.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600004503.mm.1 TC1600000475.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600004502.mm.1 NM_175256.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600004502.mm.1 NM_175256.5 Refseq
Affymetrix Mouse Clariom S Array PSR1600004502.mm.1 TC1600000475.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0