This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342497_m1
See other SCN10A GE Assays ›
Gene Symbol:
Scn10a
Gene Name:
sodium channel, voltage-gated, type X, alpha
Gene Aliases:
Nav1.8, PN3, SNS
Chromosome Location:
Chr.9: 119608456 - 119719032 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342497_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Scn10a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001205321.1 NP_001192250.1 4 - 5 539 66
NM_009134.3 NP_033160.2 4 - 5 539 66
GenBank mRNA AK135787.1 - 1 - 2 160 66
AK157866.1 - 4 - 5 539 66
AY538273.1 - 3 - 4 469 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003245.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Scn10a

Panther Classification:

Molecular Function -

voltage-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of heart rate
transport
ion transport
sodium ion transport
neuronal action potential
sensory perception of pain
regulation of ion transmembrane transport
sodium ion transmembrane transport
transmembrane transport
regulation of cardiac muscle contraction
regulation of atrial cardiac muscle cell membrane depolarization
cardiac conduction
membrane depolarization during action potential
AV node cell action potential
bundle of His cell action potential
ion channel activity
voltage-gated ion channel activity
voltage-gated sodium channel activity
sodium channel activity
ion channel binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027358.mm.1 NM_001205321.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027358.mm.1 NM_001205321.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027358.mm.1 NM_009134.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900027358.mm.1 NM_009134.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900027358.mm.1 TC0900003245.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0