This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342468_g1
See other PLCD1 GE Assays ›
Gene Symbol:
Plcd1
Gene Name:
phospholipase C, delta 1
Gene Aliases:
AW212592, C79986
Chromosome Location:
Chr.9: 119071527 - 119093502 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342468_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293648.1 NP_001280577.1 9 - 10 1606 53
NM_019676.3 NP_062650.1 9 - 10 1557 53
GenBank mRNA AF133125.1 - 9 - 10 1539 53
AK028749.1 - 9 - 10 1558 53
AK082890.1 - 9 - 10 1549 53
BC025798.1 - 9 - 10 1533 5068610 53
JN635596.1 - 9 - 10 1606 53
U85711.1 - 9 - 10 1551 53

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003233.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plcd1

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
lipid metabolic process
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of norepinephrine secretion
lipid catabolic process
positive regulation of inositol trisphosphate biosynthetic process
intracellular signal transduction
regulation of cell proliferation
response to peptide hormone
regulation of cytosolic calcium ion concentration
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
response to calcium ion
labyrinthine layer blood vessel development
regulation of phospholipase C activity
phosphatidylserine binding
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
protein binding
phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
phosphoric diester hydrolase activity
hydrolase activity
GTPase activating protein binding
metal ion binding
phosphatidic acid binding
phosphatidylinositol phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900027161.mm.1 NM_019676.3 Refseq
Affymetrix Mouse Clariom S Array PSR0900027161.mm.1 TC0900003233.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0