This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342462_g1
See other PLCD1 GE Assays ›
Gene Symbol:
Plcd1
Gene Name:
phospholipase C, delta 1
Gene Aliases:
AW212592, C79986
Chromosome Location:
Chr.9: 119071527 - 119093502 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342462_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plcd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001293648.1 NP_001280577.1 10 - 11 1765 88
NM_019676.3 NP_062650.1 10 - 11 1716 88
NR_121571.1 10 - 11 1774 88
GenBank mRNA AF133125.1 - 10 - 11 1698 88
AK028749.1 - 10 - 11 1717 88
AK082890.1 - 10 - 11 1708 88
BC015249.1 - 10 - 11 1728 4241580 88
BC025798.1 - 10 - 11 1692 5068610 88
JN635596.1 - 10 - 11 1765 88
U85711.1 - 10 - 11 1710 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Plcd1

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
lipid metabolic process
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of norepinephrine secretion
lipid catabolic process
positive regulation of inositol trisphosphate biosynthetic process
intracellular signal transduction
regulation of cell proliferation
response to peptide hormone
regulation of cytosolic calcium ion concentration
positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
response to calcium ion
labyrinthine layer blood vessel development
regulation of phospholipase C activity
phosphatidylserine binding
phosphatidylinositol phospholipase C activity
phospholipase C activity
signal transducer activity
calcium ion binding
protein binding
phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
phosphoric diester hydrolase activity
hydrolase activity
GTPase activating protein binding
metal ion binding
phosphatidic acid binding
phosphatidylinositol phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0