This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01342060_m1
See other IGFBPL1 GE Assays ›
Gene Symbol:
Igfbpl1
Gene Name:
insulin-like growth factor binding protein-like 1
Gene Aliases:
2810011G06Rik, 2810453O06Rik, IGFBPL
Chromosome Location:
Chr.4: 45809507 - 45826827 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01342060_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igfbpl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018741.2 NP_061211.1 3 - 4 698 64
GenBank mRNA AB006141.1 - 3 - 4 686 64
AK012705.1 - 3 - 4 700 64
AK013339.1 - 2 - 3 224 64
BC051960.1 - 3 - 4 668 5710646 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002587.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igfbpl1

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
insulin-like growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400021069.mm.1 NM_018741.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021069.mm.1 NM_018741.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400021069.mm.1 TC0400002587.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0