This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340728_g1
See other G3BP1 GE Assays ›
Gene Symbol:
G3bp1
Gene Name:
GTPase activating protein (SH3 domain) binding protein 1
Gene Aliases:
AI849976, B430204O07, C87777, G3bp, mKIAA4115
Chromosome Location:
Chr.11: 55469752 - 55500888 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340728_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: G3bp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013716.2 NP_038744.1 9 - 10 1016 72
GenBank mRNA AB001927.1 - 9 - 10 1016 72
AK141689.1 - 9 - 10 1083 72
AK182641.1 - 2 - 3 212 72
AK220230.1 - 9 - 10 1070 72
BC021156.1 - 9 - 10 1028 4020362 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene G3bp1

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transport
negative regulation of canonical Wnt signaling pathway
nucleotide binding
nucleic acid binding
DNA binding
RNA binding
mRNA binding
helicase activity
nuclease activity
endonuclease activity
protein binding
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0