This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340651_m1
See other TMEM245 GE Assays ›
Gene Symbol:
Tmem245
Gene Name:
transmembrane protein 245
Gene Aliases:
A630051L19Rik, AI957324, D730040F13Rik
Chromosome Location:
Chr.4: 56876003 - 56947527 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340651_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem245
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175518.5 NP_780727.3 10 - 11 1701 91
GenBank mRNA AK052826.1 - 10 - 11 1696 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002709.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmem245

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400021899.mm.1 NM_175518.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021899.mm.1 NM_175518.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400021899.mm.1 TC0400002709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021901.mm.1 NM_175518.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021901.mm.1 NM_175518.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400021901.mm.1 TC0400002709.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400021898.mm.1 NM_175518.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400021898.mm.1 NM_175518.5 Refseq
Affymetrix Mouse Clariom S Array PSR0400021898.mm.1 TC0400002709.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0