Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400021905.mm.1 |
NM_175518.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021905.mm.1 |
NM_175518.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021905.mm.1 |
TC0400002709.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021908.mm.1 |
NM_175518.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021908.mm.1 |
NM_175518.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021908.mm.1 |
TC0400002709.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021906.mm.1 |
NM_175518.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021906.mm.1 |
NM_175518.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021906.mm.1 |
TC0400002709.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400021909.mm.1 |
NM_175518.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021909.mm.1 |
NM_175518.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400021909.mm.1 |
TC0400002709.mm.1 |
MTA TC |