This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01340396_g1
See other SLC27A1 GE Assays ›
Gene Symbol:
Slc27a1
Gene Name:
solute carrier family 27 (fatty acid transporter), member 1
Gene Aliases:
FATP1, Fatp
Chromosome Location:
Chr.8: 71568676 - 71587304 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01340396_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc27a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011977.3 NP_036107.1 9 - 10 1414 62
GenBank mRNA AK040923.1 - 8 - 9 1353 62
AK137700.1 - 3 - 4 511 62
AK153133.1 - 6 - 7 1155 62
AK155185.1 - 7 - 8 1220 62
AK164798.1 - 7 - 8 1193 62
AK165571.1 - 6 - 7 1236 62
BC028937.1 - 9 - 10 1414 5362790 62
U15976.1 - 7 - 8 1209 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800000796.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc27a1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

medium-chain fatty acid transport
long-chain fatty acid metabolic process
lipid metabolic process
fatty acid metabolic process
phosphatidylethanolamine biosynthetic process
phosphatidic acid biosynthetic process
phosphatidylglycerol biosynthetic process
phosphatidylcholine biosynthetic process
phosphatidylserine biosynthetic process
phosphatidylinositol biosynthetic process
transport
lipid transport
metabolic process
response to cold
fatty acid transport
long-chain fatty acid transport
positive regulation of heat generation
cardiolipin biosynthetic process
response to insulin
adiponectin-activated signaling pathway
negative regulation of phospholipid biosynthetic process
positive regulation of protein serine/threonine kinase activity
nucleotide binding
catalytic activity
acyl-CoA ligase activity
long-chain fatty acid-CoA ligase activity
succinate-CoA ligase activity
3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
fatty acid transporter activity
fatty acid ligase activity
ligase activity
3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
benzoyl acetate-CoA ligase activity
2,4-dichlorobenzoate-CoA ligase activity
very long-chain fatty acid-CoA ligase activity
pivalate-CoA ligase activity
cyclopropanecarboxylate-CoA ligase activity
adipate-CoA ligase activity
citronellyl-CoA ligase activity
mentha-1,3-dione-CoA ligase activity
thiophene-2-carboxylate-CoA ligase activity
2,4,4-trimethylpentanoate-CoA ligase activity
cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
protein homodimerization activity
branched-chain acyl-CoA synthetase (ADP-forming) activity
aryl-CoA synthetase (ADP-forming) activity
3-hydroxypropionyl-CoA synthetase activity
perillic acid:CoA ligase (ADP-forming) activity
perillic acid:CoA ligase (AMP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
pristanate-CoA ligase activity
malonyl-CoA synthetase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800005755.mm.1 NM_011977.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800005755.mm.1 NM_011977.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800005755.mm.1 TC0800000796.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-867854cbc7-vwcfx:80/100.66.130.94:80.
git-commit: 25ccc7b1e9fd8ebf0690872e1fef11e169411d8b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0