This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01339506_m1
See other PRKRA GE Assays ›
Gene Symbol:
Prkra
Gene Name:
protein kinase, interferon inducible double stranded RNA dependent activator
Gene Aliases:
AV120107, PRK, Pact, RAX, lear
Chromosome Location:
Chr.2: 76609215 - 76648199 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01339506_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011871.2 NP_036001.1 3 - 4 458 82
GenBank mRNA AF083032.1 - 3 - 4 458 82
AK012798.1 - 3 - 4 444 82
AK155112.1 - 3 - 4 482 82
BC011311.1 - 3 - 4 466 3985758 82

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkra

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
positive regulation of cell proliferation
production of siRNA involved in RNA interference
gene silencing by RNA
pre-miRNA processing
cellular response to oxidative stress
production of miRNAs involved in gene silencing by miRNA
outer ear morphogenesis
middle ear morphogenesis
skeletal system morphogenesis
positive regulation of intrinsic apoptotic signaling pathway
RNA binding
double-stranded RNA binding
protein binding
enzyme binding
protein kinase binding
identical protein binding
protein homodimerization activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0