This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01338875_g1
See other SLC28A3 GE Assays ›
Gene Symbol:
Slc28a3
Gene Name:
solute carrier family 28 (sodium-coupled nucleoside transporter), member 3
Gene Aliases:
Cnt3
Chromosome Location:
Chr.13: 58553307 - 58610877 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01338875_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc28a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022317.3 NP_071712.3 15 - 16 1838 75
GenBank mRNA AF305211.1 - 15 - 16 1771 75
AK145077.1 - 15 - 16 1839 75
BC010472.1 - 15 - 16 1826 3586005 75
BC013783.1 - 15 - 16 1829 3600786 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002102.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc28a3

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
transport
purine nucleobase transport
pyrimidine nucleobase transport
purine nucleoside transmembrane transport
pyrimidine nucleoside transport
sodium ion transmembrane transport
pyrimidine-containing compound transmembrane transport
nucleoside transmembrane transport
nucleoside binding
nucleoside transmembrane transporter activity
nucleoside:sodium symporter activity
pyrimidine- and adenine-specific:sodium symporter activity
purine-specific nucleoside:sodium symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300013436.mm.1 NM_022317.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300013436.mm.1 NM_022317.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300013436.mm.1 TC1300002102.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0