Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200036939.mm.1 |
NM_001033136.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036939.mm.1 |
NM_001033136.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036939.mm.1 |
TC0200004423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036942.mm.1 |
NM_001033136.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036942.mm.1 |
NM_001033136.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036942.mm.1 |
TC0200004423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036934.mm.1 |
NM_001033136.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036934.mm.1 |
NM_001033136.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036934.mm.1 |
TC0200004423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036940.mm.1 |
NM_001033136.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036940.mm.1 |
NM_001033136.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036940.mm.1 |
TC0200004423.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200036937.mm.1 |
NM_001033136.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036937.mm.1 |
NM_001033136.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200036937.mm.1 |
TC0200004423.mm.1 |
MTA TC |