This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337966_m1
See other BUB1B GE Assays ›
Gene Symbol:
Bub1b
Gene Name:
budding uninhibited by benzimidazoles 1 homolog, beta (S. cerevisiae)
Gene Aliases:
AU045529, BUBR1
Chromosome Location:
Chr.2: 118598203 - 118641592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337966_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bub1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009773.3 NP_033903.2 4 - 5 605 100
GenBank mRNA AF107296.1 - 4 - 5 560 100
AK133909.1 - 4 - 5 546 100
BC031577.1 - 4 - 5 556 4953223 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001741.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bub1b

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
apoptotic process
cell cycle
mitotic nuclear division
metaphase/anaphase transition of mitotic cell cycle
phosphorylation
protein localization to kinetochore
cell division
protein localization to chromosome, centromeric region
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200012875.mm.1 NM_009773.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200012875.mm.1 NM_009773.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200012875.mm.1 TC0200001741.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0