This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01337300_g1
See other PPP1R12C GE Assays ›
Gene Symbol:
Ppp1r12c
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 12C
Gene Aliases:
2410197A17Rik, AI839747, B430101M08, Mbs85
Chromosome Location:
Chr.7: 4481519 - 4502222 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01337300_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r12c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029834.3 NP_084110.2 5 - 6 969 129
GenBank mRNA AK046570.1 - 5 - 6 939 129
AK144691.1 - 5 - 6 970 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002159.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppp1r12c

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700019622.mm.1 NM_029834.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700019622.mm.1 NM_029834.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700019622.mm.1 TC0700002159.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0