This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336658_m1
See other DENND1A GE Assays ›
Gene Symbol:
Dennd1a
Gene Name:
DENN/MADD domain containing 1A
Gene Aliases:
6030446I19Rik, connecdenn
Chromosome Location:
Chr.2: 37798990 - 38287384 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336658_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dennd1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146122.3 NP_666234.3 19 - 20 1742 69
GenBank mRNA AK137530.1 - 18 - 19 1670 69
BC027786.1 - 19 - 20 1705 5346095 69
CX563166.1 - 1 - 2 116 69
DQ448594.1 - 19 - 20 1487 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dennd1a

Gene Ontology Categories:

Function(s) Process(es)

transport
endocytosis
protein transport
regulation of Rab protein signal transduction
positive regulation of GTPase activity
synaptic vesicle endocytosis
guanyl-nucleotide exchange factor activity
protein binding
Rab guanyl-nucleotide exchange factor activity
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0