This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336475_g1
See other TMX2 GE Assays ›
Gene Symbol:
Tmx2
Gene Name:
thioredoxin-related transmembrane protein 2
Gene Aliases:
2310042M24Rik, AA589631, Txndc14
Chromosome Location:
Chr.2: 84671311 - 84678828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336475_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmx2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290751.1 NP_001277680.1 3 - 4 463 93
NM_025868.4 NP_080144.1 5 - 6 763 93
GenBank mRNA AK009759.1 - 4 - 5 451 93
AK075750.1 - 5 - 6 765 93
AK145245.1 - 4 - 5 472 93
AK170587.1 - 4 - 5 460 93
AK177742.1 - 1 - 2 54 93
BC013544.1 - 4 - 5 445 4242418 93
BC110994.1 - 4 - 5 446 5349078 93

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmx2

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

biological_process
cell redox homeostasis
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200033509.mm.1 NM_025868.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200033509.mm.1 TC0200003873.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0