This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01336353_m1
See other CAMK1D GE Assays ›
Gene Symbol:
Camk1d
Gene Name:
calcium/calmodulin-dependent protein kinase ID
Gene Aliases:
A630059D12Rik, CKLiK, CaMKIdelta, E030025C11Rik, mCKLiK
Chromosome Location:
Chr.2: 5293457 - 5715364 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01336353_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290374.1 NP_001277303.1 8 - 9 1372 61
NM_001290375.1 NP_001277304.1 9 - 10 1447 61
NM_001290376.1 NP_001277305.1 9 - 10 963 61
NM_177343.4 NP_796317.2 9 - 10 1447 61
GenBank mRNA AK052401.1 - 9 - 10 868 61
AK053173.1 - 9 - 10 1201 61
AK147616.1 - 9 - 10 1448 61
AK147657.1 - 9 - 10 1158 61
AK154434.1 - 9 - 10 1036 61
AK170123.1 - 8 - 9 1131 61
AK170777.1 - 9 - 10 1212 61
AY273822.1 - 9 - 10 963 61
BC141413.1 - 9 - 10 947 9056026 61
BC141414.1 - 9 - 10 947 9056027 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002876.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camk1d

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
inflammatory response
nervous system development
metabolic process
positive regulation of neuron projection development
phosphorylation
positive regulation of CREB transcription factor activity
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of phagocytosis
regulation of dendrite development
positive regulation of respiratory burst
regulation of granulocyte chemotaxis
positive regulation of neutrophil chemotaxis
nucleotide binding
catalytic activity
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023861.mm.1 NM_177343.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200023861.mm.1 TC0200002876.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0