This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335895_m1
See other ZIM3 GE Assays ›
Gene Symbol:
Zim3
Gene Name:
zinc finger, imprinted 3
Gene Aliases:
1700128I23Rik
Chromosome Location:
Chr.7: 6955685 - 6976662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335895_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zim3
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_036631.2
5 - 6 488 63
GenBank mRNA AF365932.1 - LIT1822
Zim3
5 - 6 493 63
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CF914807.1 - 2 - 3 284 Chr. 7 - 6963335 - 6975464 [-] on Build GRCm38 63

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 3099 Mir3099 mir-3099,
mmu-mir-3099
-157.5kb Anti-sense Chr. 7 - 6803587 - 6803670 on NCBI Build GRCm38
Downstream zinc finger protein 264 Zfp264 11.4kb Anti-sense Chr. 7 - 6977413 - 6985640 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002219.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700020151.mm.1 NR_036631.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700020151.mm.1 NR_036631.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700020151.mm.1 TC0700002219.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0