This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335793_m1
See other 4930583P06RIK GE Assays ›
Gene Symbol:
4930583P06Rik
Gene Name:
RIKEN cDNA 4930583P06 gene
Gene Aliases:
-
Chromosome Location:
Chr.2: 124217679 - 124222358 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335793_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930583P06Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040612.1 2 - 3 394 146
GenBank mRNA AK016345.1 - 2 - 3 366 146
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA465385.1 - 2 - 3 397 Chr. 2 - 124217689 - 124222360 [-] on Build GRCm38 146

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream sema domain,
transmembrane domain (TM),
and cytoplasmic domain,
(semaphorin) 6D
Sema6d 1110067B02Rik,
AA409156,
D330011G23,
mKIAA1479
Anti-sense Chr. 2 - 124089969 - 124667789 on NCBI Build GRCm38
Downstream solute carrier family 24,
member 5
Slc24a5 F630045L20Rik,
JSX,
NCKX5,
NCX5,
Oca6
848.6kb Anti-sense Chr. 2 - 125068127 - 125088677 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200004517.mm.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135873 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01913973 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595002 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01540875 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01637050 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595010 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595038 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01297897 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595061 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01971205 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595060 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01966478 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595064 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01971195 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595033 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594994 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595042 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01297900 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595037 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595017 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02043816 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01547908 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594987 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595056 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595050 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01547916 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594988 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595065 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595074 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02276974 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595054 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01297878 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595008 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595039 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595004 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595040 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594990 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595062 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595005 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01637058 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595009 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595012 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594992 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595036 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02201650 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595057 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595046 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01547913 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595049 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594986 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P01297884 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595059 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21594997 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595028 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595067 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P21595014 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200038390.mm.1 NR_040612.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038390.mm.1 NR_040612.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200038390.mm.1 TC0200004517.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 790548 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0