This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01335382_g1
See other 4930455J16RIK GE Assays ›
Gene Symbol:
4930455J16Rik
Gene Name:
RIKEN cDNA 4930455J16 gene
Gene Aliases:
-
Chromosome Location:
Chr.13: 58741074 - 58762311 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01335382_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930455J16Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK030052.1 - 5 - 6 703 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream solute carrier family 28 (sodium-coupled nucleoside transporter),
member 3
Slc28a3 Cnt3 -141.8kb Sense Chr. 13 - 58553307 - 58610877 on NCBI Build GRCm38
Downstream neurotrophic tyrosine kinase,
receptor,
type 2
Ntrk2 GP145-TrkB/GP95-TrkB,
Tkrb,
trk-B,
trkB
51.5kb Anti-sense Chr. 13 - 58806569 - 59133970 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0