Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200015386.mm.1 |
NM_001177674.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015386.mm.1 |
NM_001177674.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015386.mm.1 |
TC1200002236.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_001177673.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_001177673.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_001177674.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_001177674.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_175493.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
NM_175493.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015385.mm.1 |
TC1200002236.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015388.mm.1 |
NM_001177674.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015388.mm.1 |
NM_001177674.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015388.mm.1 |
TC1200002236.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200015387.mm.1 |
NM_001177674.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015387.mm.1 |
NM_001177674.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200015387.mm.1 |
TC1200002236.mm.1 |
MTA TC |