Assay Details
Gene Symbol: Pdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC025480.1 | - | 5 - 6 | 655 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: Pdk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC025480.1 | - | 5 - 6 | 655 | 92 |
Set Membership: |
![]() ![]() ![]() ![]() |