This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334445_g1
See other SEMA6D GE Assays ›
Gene Symbol:
Sema6d
Gene Name:
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D
Gene Aliases:
1110067B02Rik, AA409156, D330011G23, mKIAA1479
Chromosome Location:
Chr.2: 124089969 - 124667789 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334445_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema6d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290997.1 NP_001277926.1 11 - 12 1245 75
NM_001291000.1 NP_001277929.1 7 - 8 1363 75
NM_172537.4 NP_766125.2 7 - 8 1363 75
NM_199238.3 NP_954708.1 7 - 8 1363 75
NM_199239.3 NP_954709.1 6 - 7 598 75
NM_199240.3 NP_954710.1 6 - 7 598 75
NM_199241.3 NP_954711.1 6 - 7 598 75
GenBank mRNA AB091532.1 - 7 - 8 1363 75
AB091533.1 - 6 - 7 544 75
AB091534.1 - 6 - 7 544 75
AB091535.1 - 6 - 7 544 75
AB091536.1 - 6 - 7 544 75
AK031307.1 - 7 - 8 1059 75
AK052232.1 - 7 - 8 771 75
AK084922.1 - 7 - 8 1060 75
AK122515.1 - 11 - 12 1248 75
BC060680.1 - 7 - 8 657 6406252 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sema6d

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
nervous system development
positive regulation of smooth muscle cell migration
negative regulation of smooth muscle cell migration
ventricular system development
cell differentiation
protein binding
semaphorin receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0