This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334382_m1
See other CCDC73 GE Assays ›
Gene Symbol:
Ccdc73
Gene Name:
coiled-coil domain containing 73
Gene Aliases:
2210415I11Rik, 4931417A20, AV080665, BC095976
Chromosome Location:
Chr.2: 104886324 - 104999737 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334382_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ccdc73
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177600.4 NP_808268.2 7 - 8 625 149
GenBank mRNA BC095976.1 - 6 - 7 371 149

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200001572.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ccdc73

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200011776.mm.1 NM_177600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200011776.mm.1 NM_177600.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200011776.mm.1 TC0200001572.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200011775.mm.1 NM_177600.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200011775.mm.1 NM_177600.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200011775.mm.1 TC0200001572.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header