This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01334080_m1
See other CD82 GE Assays ›
Gene Symbol:
Cd82
Gene Name:
CD82 antigen
Gene Aliases:
AA682076, AL023070, C33, IA4, Kai1, Tspan27
Chromosome Location:
Chr.2: 93419102 - 93462950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01334080_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cd82
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136055.2 NP_001129527.1 2 - 3 113 120
NM_001271432.1 NP_001258361.1 3 - 4 332 120
NM_001271461.1 NP_001258390.1 2 - 3 124 120
NM_001271462.1 NP_001258391.1 2 - 3 136 120
NM_007656.5 NP_031682.1 2 - 3 245 120
GenBank mRNA AK135381.1 - 2 - 3 109 120
AK146912.1 - 2 - 3 109 120
AK152871.1 - 2 - 3 240 120
AK153070.1 - 2 - 3 240 120
AK158033.1 - 2 - 3 188 120
AK168097.1 - 2 - 3 196 120
AK168515.1 - 2 - 3 215 120
BC132559.1 - 2 - 3 215 40130836 120
BC132565.1 - 2 - 3 215 40130842 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK135381.1, AK146912.1, AK153070.1, AK168097.1, AK168515.1, BC132559.1, BC132565.1, NM_001136055.2, NM_001271432.1, NM_001271461.1, NM_001271462.1, NM_007656.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cd82

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200034898.mm.1 NM_001271432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034898.mm.1 TC0200004159.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001136055.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001271430.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001271431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001271432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001271461.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_001271462.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 NM_007656.5 Refseq
Affymetrix Mouse Clariom S Array PSR0200034896.mm.1 TC0200004159.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200034900.mm.1 NM_001271462.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034900.mm.1 TC0200004159.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200034913.mm.1 NM_001136055.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200034913.mm.1 NM_001271431.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034913.mm.1 TC0200004159.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200034903.mm.1 NM_001271461.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200034903.mm.1 TC0200004159.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0