This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333998_g1
See other SSFA2 GE Assays ›
Gene Symbol:
Ssfa2
Gene Name:
sperm specific antigen 2
Gene Aliases:
5730488C15Rik, CS-1, CS1, KRAP, SPAG13, mKIAA1927
Chromosome Location:
Chr.2: 79635373 - 79672966 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333998_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ssfa2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080558.4 NP_542125.3 2 - 3 365 73
GenBank mRNA AB093303.1 - 2 - 3 240 73
AB120565.1 - 2 - 3 250 73
AK169543.1 - 2 - 3 366 73
BC122519.1 - 2 - 3 251 73
BC141883.1 - 2 - 3 250 40097831 73
CF539296.1 - 2 - 3 181 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ssfa2

Gene Ontology Categories:

Process(es)

actin binding
receptor binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200009345.mm.1 NM_080558.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200009345.mm.1 NM_080558.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200009345.mm.1 TC0200001164.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0