This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333164_m1
See other HSPA14 GE Assays ›
Gene Symbol:
Hspa14
Gene Name:
heat shock protein 14
Gene Aliases:
70kDa, HSP70L1, Hsp70-4, NST-1, hsr.1
Chromosome Location:
Chr.2: 3488849 - 3512936 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333164_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hspa14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_015765.2 NP_056580.2 10 - 11 1120 73
GenBank mRNA AK031489.1 - 1059 73
AK145937.1 - 10 - 11 1074 73
AK153397.1 - 10 - 11 1082 73
BC002056.1 - 10 - 11 1048 3493119 73
BI734146.1 - 2 - 3 223 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002851.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hspa14

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
ATP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200023666.mm.1 NM_015765.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200023666.mm.1 NM_015765.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200023666.mm.1 TC0200002851.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0