This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01333112_g1
See other RNF44 GE Assays ›
Gene Symbol:
Rnf44
Gene Name:
ring finger protein 44
Gene Aliases:
AI854545, mKIAA1100
Chromosome Location:
Chr.13: 54679399 - 54693995 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01333112_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rnf44
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146025.1 NP_001139497.1 6 - 7 1162 81
NM_001146026.1 NP_001139498.1 6 - 7 1087 81
NM_001146027.1 NP_001139499.1 6 - 7 953 81
NM_134064.2 NP_598825.2 6 - 7 1095 81
NR_027395.1 6 - 7 1015 81
NR_027396.1 6 - 7 1012 81
GenBank mRNA AK147349.1 - 6 - 7 1088 81
AK160090.1 - 6 - 7 946 81
AK160115.1 - 6 - 7 946 81
AK161651.1 - 6 - 7 1075 81
AK171943.1 - 6 - 7 944 81
BC017630.1 - 6 - 7 888 4507289 81
BC035548.1 - 6 - 7 910 4221284 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002038.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rnf44

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146025.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146025.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146026.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146026.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146027.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_001146027.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_134064.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NM_134064.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NR_027395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NR_027395.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NR_027396.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 NR_027396.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012751.mm.1 TC1300002038.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146025.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146025.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146026.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146026.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146027.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_001146027.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_134064.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NM_134064.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NR_027395.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NR_027395.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NR_027396.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 NR_027396.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300012750.mm.1 TC1300002038.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0